Learn More
Waters Corp Symmetry C18 Column, 100 Å, 5 μm, 4.6 mm × 250 mm
Catalog No. 502391778 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is equipped with an electronic eConnect column tag that contains unique column-specific information. These NFC-enabled device-tagged columns were developed for automatic identification, tracking, and digital tracing of column use history. eConnect columns can be used on any HPLC system, but their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
  • Equipped with an electronic eConnect column tag for automatic identification and tracking.
  • Features 100 Å pore size, 5 μm particle size, and 4.6 mm inner diameter × 250 mm length.
  • C18 chemistry with reversed phase separation mode.
  • Silica particle substrate, endcapped with spherical shape, and low silanol activity.
  • Operates within a pH range of 2 pH to 8 pH and supports a maximum pressure of 6000 psi (415 Bar).
  • Suitable for small molecule HPLC applications.
Catalog No. 50-239-1778 Supplier Waters Corp Supplier No. WAT054275RF
This column is equipped with an electronic eConnect column tag that contains unique column-specific information. These NFC-enabled device-tagged columns were developed for automatic identification, tracking, and digital tracing of column use history. eConnect columns can be used on any HPLC system, but their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
  • Equipped with an electronic eConnect column tag for automatic identification and tracking.
  • Features 100 Å pore size, 5 μm particle size, and 4.6 mm inner diameter × 250 mm length.
  • C18 chemistry with reversed phase separation mode.
  • Silica particle substrate, endcapped with spherical shape, and low silanol activity.
  • Operates within a pH range of 2 pH to 8 pH and supports a maximum pressure of 6000 psi (415 Bar).
  • Suitable for small molecule HPLC applications.