Learn More
Waters Corp Symmetry Column, C18, 100Å, 3.5 μm, 4.6 mm × 50 mm
Supplier:  Waters Corp WAT200625
Encompass_Preferred
These C18 columns feature monofunctionally bonded C18 ligands on high purity base-deactivated silica. They provide highly reproducible chromatographic results due to tight specifications and narrow ligand surface coverage, minimizing resolution shifts and ensuring consistent batch-to-batch performance. This design eliminates continuous method revalidation.
  • Monofunctionally bonded C18 ligands on high purity base-deactivated silica.
  • Tight column specifications ensure highly reproducible chromatography.
  • Narrow ligand surface coverage minimizes resolution shifts and ensures consistent batch results.
  • Manufactured with high purity raw materials and precise packing procedures.
  • Includes a three-page Certificate of Analysis with 22 critical quality test results.
  • Reduces need for continuous revalidation and method redevelopment.
  • Contributes to labor cost savings.
Catalog No. 50-786-108