Learn More
Waters Corp Waters Corporation Column, C18, 5 μm, 2.1 mm X 150 mm, 100 A
Catalog No. 50785834 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C18 columns are designed with monofunctionally bonded C18 ligands on high purity base-deactivated silica, ensuring highly reproducible chromatographic results. This design maximizes analytical efficiency, precision, and unsurpassed reproducibility.
  • Monofunctionally bonded C18 ligands on high purity base-deactivated silica ensure highly reproducible chromatographic results.
  • Industry’s tightest ligand surface coverage standards reduce variations in resolution and ensure consistent batch–to–batch results.
  • Designed to simplify achieving exceptional repeatability, reducing the need for revalidation and redeveloping procedures.
  • Provided with a three–page Certificate of Analysis detailing specifications and results from 22 quality tests.
  • Manufactured in cGMP, ISO9001, and ISO13485–certified facilities.
Catalog No. 50-785-834 Supplier Waters Corp Supplier No. WAT056975
Symmetry C18 columns are designed with monofunctionally bonded C18 ligands on high purity base-deactivated silica, ensuring highly reproducible chromatographic results. This design maximizes analytical efficiency, precision, and unsurpassed reproducibility.
  • Monofunctionally bonded C18 ligands on high purity base-deactivated silica ensure highly reproducible chromatographic results.
  • Industry’s tightest ligand surface coverage standards reduce variations in resolution and ensure consistent batch–to–batch results.
  • Designed to simplify achieving exceptional repeatability, reducing the need for revalidation and redeveloping procedures.
  • Provided with a three–page Certificate of Analysis detailing specifications and results from 22 quality tests.
  • Manufactured in cGMP, ISO9001, and ISO13485–certified facilities.