Learn More
Waters Corp Waters Symmetry C18, 100Å, 5 μm, 3 mm × 250 mm
Catalog No. 50780456 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C18 columns, made with high purity base–deactivated silica and monofunctionally bonded C18 ligands, offer highly reproducible chromatographic results and are manufactured using specialized packing techniques for reliable HPLC applications.
  • Highly reproducible chromatographic results due to tight column specifications.
  • Exceptional reproducibility with minimal resolution variations and repeatable batch–to–batch results.
  • Built with high purity raw materials and specialized column packing techniques.
  • Includes a 3–page Certificate of Analysis, detailing 22 quality certification tests.
  • Eliminates revalidation, reducing labor and lab expenditures.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities with rigorous monitoring.
Catalog No. 50-780-456 Supplier Waters Corp Supplier No. 186000690
Symmetry C18 columns, made with high purity base–deactivated silica and monofunctionally bonded C18 ligands, offer highly reproducible chromatographic results and are manufactured using specialized packing techniques for reliable HPLC applications.
  • Highly reproducible chromatographic results due to tight column specifications.
  • Exceptional reproducibility with minimal resolution variations and repeatable batch–to–batch results.
  • Built with high purity raw materials and specialized column packing techniques.
  • Includes a 3–page Certificate of Analysis, detailing 22 quality certification tests.
  • Eliminates revalidation, reducing labor and lab expenditures.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities with rigorous monitoring.