Learn More
Waters Corp Waters Column, C18, 100 Å, 5 μm, 4.6 mm × 250 mm
Supplier:  Waters Corp WAT054275
Encompass_Preferred
Symmetry columns are designed for HPLC drug assays, offering reliable long-term compliance. They ensure consistent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility. This specific HPLC column is a silica-based C18 column, suitable for analytical challenges and supporting successful MS adoption. It is also suitable for both analytical and preparative chromatography applications.
  • Sets the standard for HPLC drug assays, ensuring long-term compliance.
  • Provides confidence in peak shape, selectivity, column-to-column, and batch-to-batch reproducibility.
  • Silica-based HPLC column designed for particle separation speed and quality.
  • Options for column protection are available, such as protection cartridges, to extend column lifetime and remove contaminants.
  • Suitable for both analytical and preparative chromatography applications.
Catalog No. 50-785-708