Learn More
Waters Corp Waters Symmetry C18 Column, 100Å, 5 μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186002616
Encompass_Preferred
  • Maximize analytical efficiency and precision.
  • Monofunctionally bonded C18 ligand and high purity base-deactivated silica provide unmatched reproducibility.
  • Narrow ligand surface coverage specifications in the industry, leading to minimal shifts in resolution and more reproducible batch-to-batch results.
  • Achieve outstanding reproducibility year on year, eliminating the need to continuously revalidate and redevelop methods with each new batch of columns.
  • Manufactured at cGMP, ISO9001, and ISO13485 certified facilities, ensuring strict conditions and consistent results.
  • Engineered with high purity raw materials and precise column packing procedures for highly reproducible HPLC columns.
  • Delivered with a three-page Certificate of Analysis, detailing specifications and results of 22 critical quality assurance tests.
Catalog No. 50-781-245