Learn More
Waters Corp Symmetry C18 Method Validation Kit, 100Å, 3.5 µm, 4.6 mm X 150 mm, 3/pk
Catalog No. 50785996 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit consists of three Symmetry C18 columns packed with three different batches of sorbent.
Catalog No. 50-785-996 Supplier Waters Corp Supplier No. WAT094240
This kit consists of three Symmetry C18 columns packed with three different batches of sorbent.