Learn More
Waters Corp Waters Symmetry Method Validation Kit, C18, 5 μm, 4.6 mm X 250 mm
Catalog No. 50785726 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit consists of three C18 columns packed with different batches of sorbent, ensuring unparalleled reproducibility. It uses high purity base-deactivated silica and a monofunctional bound C18 ligand, resulting in consistent batch-to-batch results and reduced variations in resolution.
  • Utilizes high purity base-deactivated silica and a monofunctional bound C18 ligand.
  • Ensures unparalleled reproducibility and consistent batch-to-batch results.
  • Adheres to the industry’s tightest ligand surface coverage standards.
  • Saves labor by eliminating constant method validation.
  • Manufactured with high-purity raw materials for repeatable HPLC columns.
  • Includes a three-page Certificate of Analysis detailing specifications and 22 quality tests.
Catalog No. 50-785-726 Supplier Waters Corp Supplier No. WAT054450
This kit consists of three C18 columns packed with different batches of sorbent, ensuring unparalleled reproducibility. It uses high purity base-deactivated silica and a monofunctional bound C18 ligand, resulting in consistent batch-to-batch results and reduced variations in resolution.
  • Utilizes high purity base-deactivated silica and a monofunctional bound C18 ligand.
  • Ensures unparalleled reproducibility and consistent batch-to-batch results.
  • Adheres to the industry’s tightest ligand surface coverage standards.
  • Saves labor by eliminating constant method validation.
  • Manufactured with high-purity raw materials for repeatable HPLC columns.
  • Includes a three-page Certificate of Analysis detailing specifications and 22 quality tests.