Learn More
Waters Corp Symmetry C18 Column, 100Å, 5 μm, 3.9 mm × 20 mm
Supplier:  Waters Corp 186002086
Encompass_Preferred
These C18 Intelligent Speed (IS) Columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica. They are known for having the tightest column specifications in the industry, which leads to highly reproducible chromatographic results.
  • Monofunctionally bonded C18 ligands on high purity base-deactivated silica
  • Tightest column specifications in the industry for highly reproducible chromatographic results
Catalog No. 50-780-942