Learn More
Waters Corp VanGuard Cartridge, 300Å, 3.5 μm, 2.1 mm X 5 mm
Catalog No. 502391361 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by filtering out particulate contamination from the mobile phase stream.
  • Extends the lifetime and performance of analytical columns.
  • Filters out particulate contamination from the mobile phase stream.
  • Optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns.
  • Contains 3.5 μm sorbent particles.
Catalog No. 50-239-1361 Supplier Waters Corp Supplier No. 186007709
Symmetry C18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by filtering out particulate contamination from the mobile phase stream.
  • Extends the lifetime and performance of analytical columns.
  • Filters out particulate contamination from the mobile phase stream.
  • Optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns.
  • Contains 3.5 μm sorbent particles.