Learn More
Waters Corp Symmetry C18 VanGuard Cartridge, 3.5 μm, 3.9 mm X 5 mm, 300 Å
Catalog No. 502392194 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 3.9 mm and 4.6 mm I.D. analytical columns containing 3.5 μm sorbent particles.
  • Extends analytical column lifetime and performance by removing particulate contamination from the mobile phase stream.
  • Optimized to protect all 3.9 mm and 4.6 mm I.D. analytical columns containing 3.5 μm sorbent particles.
Catalog No. 50-239-2194 Supplier Waters Corp Supplier No. 186007711
These C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 3.9 mm and 4.6 mm I.D. analytical columns containing 3.5 μm sorbent particles.
  • Extends analytical column lifetime and performance by removing particulate contamination from the mobile phase stream.
  • Optimized to protect all 3.9 mm and 4.6 mm I.D. analytical columns containing 3.5 μm sorbent particles.