Learn More
Waters Corp Symmetry C18 VanGuard Cartridge, 300A, 5 um, 3.9 mm X 5 mm
Catalog No. 502392937 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Symmetry C18 VanGuard Cartridge extends analytical column lifetime by removing particulate contamination from the mobile phase stream. It ensures high repeatability in chromatographic results and long-term compliance for HPLC methods. Manufactured in cGMP, ISO9001, and ISO13485 certified facilities, each cartridge includes a three-page Certificate of Analysis.
  • Extends analytical column lifetime
  • Removes particulate contamination
  • Ensures high repeatability in chromatographic results
  • Supports long-term compliance of HPLC methods
  • Spherical particle shape
Catalog No. 50-239-2937 Supplier Waters Corp Supplier No. 186007715
The Symmetry C18 VanGuard Cartridge extends analytical column lifetime by removing particulate contamination from the mobile phase stream. It ensures high repeatability in chromatographic results and long-term compliance for HPLC methods. Manufactured in cGMP, ISO9001, and ISO13485 certified facilities, each cartridge includes a three-page Certificate of Analysis.
  • Extends analytical column lifetime
  • Removes particulate contamination
  • Ensures high repeatability in chromatographic results
  • Supports long-term compliance of HPLC methods
  • Spherical particle shape