Learn More
Waters Corp Waters VanGuard Cartridge, C18, 100Å, 3.5 μm, 3.9 mm X 5 mm
Catalog No. 502038176 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate and chemical contaminants from the mobile phase. Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 3.5 μm sorbent particles, they ensure compatibility by using the same packing material as C18 columns.
  • Extends column lifetime by preventing contamination.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. columns with 3.5 μm sorbent particles.
  • Uses same packing material as C18 columns for compatibility.
  • Provides consistent, high-performance protection.
  • Suitable for biological fluid cleanup and temperature management.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Batch tested for repeatable performance with various analytes.
  • Equipment tested for optimal function, improving selectivity, sensitivity, and speed.
Catalog No. 50-203-8176 Supplier Waters Corp Supplier No. 186007727
Symmetry C18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate and chemical contaminants from the mobile phase. Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 3.5 μm sorbent particles, they ensure compatibility by using the same packing material as C18 columns.
  • Extends column lifetime by preventing contamination.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. columns with 3.5 μm sorbent particles.
  • Uses same packing material as C18 columns for compatibility.
  • Provides consistent, high-performance protection.
  • Suitable for biological fluid cleanup and temperature management.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Batch tested for repeatable performance with various analytes.
  • Equipment tested for optimal function, improving selectivity, sensitivity, and speed.