Learn More
Waters Corp Waters VanGuard Cartridge, C18, 100Å, 5 μm, 3.9 mm × 5 mm
Supplier:  Waters Corp 186007731
Encompass_Preferred
This VanGuard Cartridge is designed to extend the lifespan and improve the performance of analytical columns. It achieves this by eliminating particulate contamination from the mobile phase stream.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 3.9 mm and 4.6 mm I.D. analytical columns that utilize 5 μm sorbent particles.
Catalog No. 50-153-4801