Learn More
Waters Corp VanGuard Cartridge, C4, 300 Å, 3.5 μm, 2.1 mm X 5 mm
Supplier:  Waters Corp 186007717
Encompass_Preferred
These VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns containing 3.5 μm sorbent particles.
  • Extends analytical column lifetime and performance by removing particulate contamination from the mobile phase stream.
  • Optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns containing 3.5 μm sorbent particles.
Catalog No. 50-239-1354