Learn More
Waters Corp Symmetry C4 VanGuard Cartridge, 300Ã…, 5 µm, 2.1 mm X 5 mm, 1K - 150K, 3/pk;
Catalog No. 502038174 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C4 VanGuard Cartridge, 300Å, 5 µm, 2.1 mm X 5 mm, 1K - 150K, 3/pk;
Catalog No. 50-203-8174 Supplier Waters Corp Supplier No. 186007721
Symmetry C4 VanGuard Cartridge, 300Å, 5 µm, 2.1 mm X 5 mm, 1K - 150K, 3/pk;