Learn More
Waters Corp Waters Sentry Guard Cartridge, C8, 3.5 μm, 2.1 mm × 10 mm
Catalog No. 50819046 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactivated silica. These columns have the tightest column specifications in the industry, resulting in highly reproducible chromatographic results.
  • Columns feature monofunctionally bonded C8 ligands on a high purity base-deactivated silica.
  • Columns have the tightest column specifications in the industry, giving highly reproducible chromatographic results.
Catalog No. 50-819-046 Supplier Waters Corp Supplier No. WAT106128
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactivated silica. These columns have the tightest column specifications in the industry, resulting in highly reproducible chromatographic results.
  • Columns feature monofunctionally bonded C8 ligands on a high purity base-deactivated silica.
  • Columns have the tightest column specifications in the industry, giving highly reproducible chromatographic results.