Learn More
Waters Corp Symmetry C8 Cartridge, 100Å, 3.5 µm, 4.6 mm X 100 mm, 1/pk
Supplier:  Waters Corp WAT066215
Encompass_Preferred
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results. This product needs to be used in conjunction with other parts. Please see the section below on related parts for more information.
Catalog No. 50-785-868