Learn More
Waters Corp Column, C8, 5 μm, 4.6 mm x 100 mm, 100 Å
Catalog No. 502392057 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is equipped with an electronic eConnect column tag that contains unique, column-specific information. These NFC-enabled device-tagged columns were developed to automatically identify, track, and digitally trace column use history. eConnect columns can be used on any HPLC, but their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped design
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 100 Å pore size
  • Surface area of 335
  • L7 USP classification
  • 4.6 mm inner diameter
  • 100 mm length
  • Small molecule application
Catalog No. 50-239-2057 Supplier Waters Corp Supplier No. 186002617RF
Add to Cart
Add to Cart
This column is equipped with an electronic eConnect column tag that contains unique, column-specific information. These NFC-enabled device-tagged columns were developed to automatically identify, track, and digitally trace column use history. eConnect columns can be used on any HPLC, but their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped design
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 100 Å pore size
  • Surface area of 335
  • L7 USP classification
  • 4.6 mm inner diameter
  • 100 mm length
  • Small molecule application