Learn More
Waters Corp Symmetry C8 Column, 100Å, 3.5 µm, 2.1 mm X 30 mm, 1/pk
Supplier:  Waters Corp WAT058977
Encompass_Preferred
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
Catalog No. 50-785-860