Learn More
Waters Corp Waters Chromatography Column, C8, 100Å, 3.5 μm, 4.6 x 50 mm
Supplier:  Waters Corp WAT200620
Encompass_Preferred
Symmetry C8 columns utilize monofunctionally bonded C8 ligands on a high-purity, base-deactivated silica, offering highly reproducible chromatographic results due to the industry's tightest column specifications. These columns are engineered for long life and consistent analytical techniques, providing dependable performance through the synergy of mechanical stability of the packed bed and hydraulic stability of the packing material.
  • Monofunctionally bonded C8 ligands on high purity, base-deactivated silica.
  • Highly reproducible results due to tight column specifications.
  • Extended column life, over 10,000 injections with minimal efficiency loss.
  • Superior peak shapes at neutral or low pH.
  • Manufactured to high standards with strict quality control.
  • Compatible with C8 VanGuard Cartridges for improved accuracy.
Catalog No. 50-786-106