Learn More
Waters Corp Waters Symmetry C8 Column, 100Å, 5 μm, 3 mm, 150 mm
Catalog No. 50785699 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Composed of monofunctional bonded C8 ligands on a high-purity base-deactivated silica.
  • Toughest column standards in the industry.
  • Remarkable repeatability, eliminating the need for method revalidation with every new batch of columns.
  • Utilizes hydraulic stability of the packing material and mechanical stability of the packed bed for trustworthy performance.
  • Packing structure enables improved peak shapes due to high bonding density and low residual silanol activity.
  • Retains top performance at neutral or low pH, optimizing methods.
  • Made in accordance with the strictest quality control requirements.
  • Provides long life in established analytical procedures.
  • Lasts over 10,000 injections with almost no efficiency loss, minimal increase in backpressure, and minimal change in retention time.
Catalog No. 50-785-699 Supplier Waters Corp Supplier No. WAT054230
  • Composed of monofunctional bonded C8 ligands on a high-purity base-deactivated silica.
  • Toughest column standards in the industry.
  • Remarkable repeatability, eliminating the need for method revalidation with every new batch of columns.
  • Utilizes hydraulic stability of the packing material and mechanical stability of the packed bed for trustworthy performance.
  • Packing structure enables improved peak shapes due to high bonding density and low residual silanol activity.
  • Retains top performance at neutral or low pH, optimizing methods.
  • Made in accordance with the strictest quality control requirements.
  • Provides long life in established analytical procedures.
  • Lasts over 10,000 injections with almost no efficiency loss, minimal increase in backpressure, and minimal change in retention time.