Learn More
Waters Corp Waters Column, C8, 100 Å, 5 μm, 3.9 mm X 150 mm
Catalog No. 50785641 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C8 columns use monofunctionally bonded C8 ligands on high purity base-deactivated silica, offering highly reproducible chromatographic results. They provide long life for analytical methods, lasting over 10,000 injections with minimal loss in efficiency, backpressure increase, or retention time change.
  • Tightest column specifications in the industry.
  • Offers highly reproducible chromatographic results and extended method life.
  • Maintains minimal loss in efficiency, backpressure, and retention time after 10,000+ injections.
  • Ensures consistent performance through optimized hydraulic and mechanical stability.
  • Achieves superior peak shape across pH 2–8 due to high bonding density and low silanol activity.
  • Allows method optimization at neutral or low pH without compromising peak shape.
Catalog No. 50-785-641 Supplier Waters Corp Supplier No. WAT046970
Symmetry C8 columns use monofunctionally bonded C8 ligands on high purity base-deactivated silica, offering highly reproducible chromatographic results. They provide long life for analytical methods, lasting over 10,000 injections with minimal loss in efficiency, backpressure increase, or retention time change.
  • Tightest column specifications in the industry.
  • Offers highly reproducible chromatographic results and extended method life.
  • Maintains minimal loss in efficiency, backpressure, and retention time after 10,000+ injections.
  • Ensures consistent performance through optimized hydraulic and mechanical stability.
  • Achieves superior peak shape across pH 2–8 due to high bonding density and low silanol activity.
  • Allows method optimization at neutral or low pH without compromising peak shape.