Learn More
Waters Corp Waters Sentry Guard Cartridge, C8, 5 μm, 3.9 mm, 20 mm
Catalog No. 50785703 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C8 columns utilize monofunctionally bonded C8 ligands on high purity base-deactivated silica for highly reproducible chromatographic results. This Sentry Guard Cartridge extends column longevity by preventing particles and contaminants from reaching the analytical column. It employs high-performance packing materials, manufactured in a cGMP, ISO 9001-certified facility, with rigorous testing ensuring consistent performance.
  • Highly reproducible chromatographic results.
  • Extends analytical column life.
  • Prevents particles and chemical contaminants from analytical column.
  • Uses high-performance packing materials.
  • Designed for biological fluid cleansing and temperature control.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Packing material chromatographically tested for consistent performance.
Catalog No. 50-785-703 Supplier Waters Corp Supplier No. WAT054250
Symmetry C8 columns utilize monofunctionally bonded C8 ligands on high purity base-deactivated silica for highly reproducible chromatographic results. This Sentry Guard Cartridge extends column longevity by preventing particles and contaminants from reaching the analytical column. It employs high-performance packing materials, manufactured in a cGMP, ISO 9001-certified facility, with rigorous testing ensuring consistent performance.
  • Highly reproducible chromatographic results.
  • Extends analytical column life.
  • Prevents particles and chemical contaminants from analytical column.
  • Uses high-performance packing materials.
  • Designed for biological fluid cleansing and temperature control.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Packing material chromatographically tested for consistent performance.