Learn More
Waters Corp Symmetry C8 Intelligent Speed (IS) Column, 100Å, 3.5 µm, 3.9 mm X 20 mm, 1/pk
Catalog No. 50780939 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
Catalog No. 50-780-939 Supplier Waters Corp Supplier No. 186002083
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.