Learn More
Waters Corp VanGuard Cartridge, 100Å, 3.5 μm, 3.9 mm x 5 mm
Supplier:  Waters Corp 186007735
Encompass_Preferred
Symmetry C8 VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns containing 3.5 μm sorbent particles.
  • Extends analytical column lifetime without compromising chromatographic performance.
  • Removes particulates and chemical contamination to prevent degradation of column performance.
  • Helps maintain the separation efficiency of UPLC™, UHPLC, and HPLC systems.
  • Offers a cost-effective solution for protecting analytical columns.
  • Compatible with analytical columns having internal diameters of 2.1 mm, 3.0 mm, 3.9 mm, and 4.6 mm.
  • Available in a variety of particle sizes and stationary phases for physical and chemical protection.
Catalog No. 50-239-1524