Learn More
Waters Corp VanGuard Cartridge, C8, 100Å, 5μm, 3.9 mm x 5 mm
Catalog No. 502391698 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C8 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase. These cartridges protect 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent particles, positioned between the injector and the analytical column.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns containing 5 μm sorbent particles.
  • Improves analytical column efficiency by blocking chemical impurities and suspended particles.
  • Constructed from the same high-performance packing materials as analytical columns for consistent performance.
  • Manufactured in a cGMP, ISO 9001-certified facility using ultra-pure reagents, ensuring quality and durability.
  • Each batch is chromatographically tested to high standards, with equipment tested for peak performance, selectivity, and sensitivity.
Catalog No. 50-239-1698 Supplier Waters Corp Supplier No. 186007739
Symmetry C8 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase. These cartridges protect 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent particles, positioned between the injector and the analytical column.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns containing 5 μm sorbent particles.
  • Improves analytical column efficiency by blocking chemical impurities and suspended particles.
  • Constructed from the same high-performance packing materials as analytical columns for consistent performance.
  • Manufactured in a cGMP, ISO 9001-certified facility using ultra-pure reagents, ensuring quality and durability.
  • Each batch is chromatographically tested to high standards, with equipment tested for peak performance, selectivity, and sensitivity.