Learn More
Waters Corp Symmetry Shield RP18 VanGuard Cartridge, 100Å, 3.5 μm, 2.1 mm x 5 mm
Catalog No. 502390600 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry Shield RP18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream.
  • Designed to protect all 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles.
Catalog No. 50-239-0600 Supplier Waters Corp Supplier No. 186007749
Symmetry Shield RP18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream.
  • Designed to protect all 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles.