Learn More
Waters Corp VanGuard Cartridge, Symmetry Shield RP18, 100Å, 5 μm
Catalog No. 502391253 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SymmetryShield RP18 VanGuard Cartridges prolong analytical column lifetime and enhance performance. They filter particulate contamination and block chemical impurities from the mobile phase stream, offering protection for 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Extends analytical column lifespan and enhances performance by removing particulate contamination and blocking chemical impurities.
  • Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Functions as a protective barrier between the injector and analytical column.
  • Built with high-performance packing materials.
  • Designed for biological fluid cleanup and temperature control.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Undergoes rigorous chromatographic testing and unit testing for reliable performance and selectivity.
Catalog No. 50-239-1253 Supplier Waters Corp Supplier No. 186007755
SymmetryShield RP18 VanGuard Cartridges prolong analytical column lifetime and enhance performance. They filter particulate contamination and block chemical impurities from the mobile phase stream, offering protection for 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Extends analytical column lifespan and enhances performance by removing particulate contamination and blocking chemical impurities.
  • Optimized for 3.9 mm and 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Functions as a protective barrier between the injector and analytical column.
  • Built with high-performance packing materials.
  • Designed for biological fluid cleanup and temperature control.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Undergoes rigorous chromatographic testing and unit testing for reliable performance and selectivity.