Learn More
Waters Corp VanGuard Cartridge, Symmetry Shield RP18, 100Å, 5 μm, 2.1 mm x 5 mm
Catalog No. 502392138 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry Shield RP18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns. These cartridges remove particulate contamination from the mobile phase stream.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns.
  • Compatible with columns containing 5 μm sorbent particles.
Catalog No. 50-239-2138 Supplier Waters Corp Supplier No. 186007753
Symmetry Shield RP18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns. These cartridges remove particulate contamination from the mobile phase stream.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns.
  • Compatible with columns containing 5 μm sorbent particles.