Learn More
Waters Corp VanGuard Cartridge, 100Å, 3.5 μm, 3.9 mm x 5 mm
Catalog No. 502392162 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Symmetry Shield RP18 VanGuard Cartridge is designed to extend the lifetime and performance of analytical columns by efficiently removing particulate contamination from the mobile phase stream.
  • Extends analytical column lifetime and performance by removing particulate contamination.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns containing 3.5 μm sorbent particles.
  • Constructed from the same high-performance packing materials as analytical columns.
  • Dependable for increased capacity in biological fluid cleanup and temperature control.
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure chemicals.
  • Each batch of packing material is chromatographically tested with acidic, basic, and neutral analytes.
Catalog No. 50-239-2162 Supplier Waters Corp Supplier No. 186007751
The Symmetry Shield RP18 VanGuard Cartridge is designed to extend the lifetime and performance of analytical columns by efficiently removing particulate contamination from the mobile phase stream.
  • Extends analytical column lifetime and performance by removing particulate contamination.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns containing 3.5 μm sorbent particles.
  • Constructed from the same high-performance packing materials as analytical columns.
  • Dependable for increased capacity in biological fluid cleanup and temperature control.
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure chemicals.
  • Each batch of packing material is chromatographically tested with acidic, basic, and neutral analytes.