Learn More
Waters Corp VanGuard Cartridge, Symmetry Shield RP8, 100Å, 5 μm, 2.1 mm × 5 mm
Catalog No. 502391605 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard Cartridge extends analytical column lifetime and performance by removing particulate and chemical contamination from the mobile phase. It is compatible with 2.1 mm and 3.0 mm I.D. Symmetry Shield RP8 analytical columns utilizing 5 μm sorbent particles.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Provides cost-effective protection for analytical columns.
  • Compatible with 2.1 mm and 3.0 mm I.D. analytical columns.
  • Features C8 chemistry with 100Å pore size.
  • Utilizes 5 μm spherical silica particles.
  • Designed for a pH range of 2 to 8.
  • Supports HPLC systems with a maximum pressure of 18000 psi (1240 bar).
Catalog No. 50-239-1605 Supplier Waters Corp Supplier No. 186007745
This VanGuard Cartridge extends analytical column lifetime and performance by removing particulate and chemical contamination from the mobile phase. It is compatible with 2.1 mm and 3.0 mm I.D. Symmetry Shield RP8 analytical columns utilizing 5 μm sorbent particles.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Provides cost-effective protection for analytical columns.
  • Compatible with 2.1 mm and 3.0 mm I.D. analytical columns.
  • Features C8 chemistry with 100Å pore size.
  • Utilizes 5 μm spherical silica particles.
  • Designed for a pH range of 2 to 8.
  • Supports HPLC systems with a maximum pressure of 18000 psi (1240 bar).