Learn More
Waters Corp Waters Corporation Column, C18, 300 Å Pore, 3.5 μm Particle, 2.1 mm × 100 mm
Supplier:  Waters Corp 186000188
Encompass_Preferred
These C18 columns are engineered for high-performance liquid chromatography (HPLC) methods, ensuring long-term compliance and high repeatability. They feature a monofunctional bound C18 ligand with high purity base-deactivated Silica, providing excellent reproducibility due to tight ligand surface coverage standards. Produced in cGMP, ISO9001, and ISO13485 certified facilities, each column is rigorously tested to meet stringent quality specifications.
  • Provides confidence in long-term compliance for HPLC methods.
  • Ensures high repeatability in chromatographic findings.
  • Offers excellent reproducibility year after year with tight ligand coverage.
  • Reduces labor and lab expenditures by minimizing revalidation needs.
  • Manufactured under strict cGMP, ISO9001, and ISO13485 certified conditions.
Catalog No. 50-780-114