Learn More
Waters Corp Waters Symmetry C18 Column, 3.5 μm, 4.6 mm, 100 mm
Catalog No. 50780116 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry columns ensure long-term compliance of HPLC methods with consistent peak shape, selectivity, and reproducibility. This C18 column achieves high reproducibility using a monofunctional bound C18 ligand and high-purity base-deactivated silica, along with tight ligand surface coverage standards for consistent batch results.
  • Unsurpassed reproducibility from monofunctional bound C18 ligand and high-purity base-deactivated silica.
  • Tightest ligand surface coverage standards, for fewer resolution shifts and consistent batch results.
  • Exceptional reproducibility year after year.
  • Reduces the need to develop and validate new methods, saving labor.
  • Produced with high-purity raw materials and advanced column packaging for repeatable HPLC performance.
  • Includes a three-page Certificate of Analysis with 22 crucial test results.
Catalog No. 50-780-116 Supplier Waters Corp Supplier No. 186000190
Add to Cart
Add to Cart
Symmetry columns ensure long-term compliance of HPLC methods with consistent peak shape, selectivity, and reproducibility. This C18 column achieves high reproducibility using a monofunctional bound C18 ligand and high-purity base-deactivated silica, along with tight ligand surface coverage standards for consistent batch results.
  • Unsurpassed reproducibility from monofunctional bound C18 ligand and high-purity base-deactivated silica.
  • Tightest ligand surface coverage standards, for fewer resolution shifts and consistent batch results.
  • Exceptional reproducibility year after year.
  • Reduces the need to develop and validate new methods, saving labor.
  • Produced with high-purity raw materials and advanced column packaging for repeatable HPLC performance.
  • Includes a three-page Certificate of Analysis with 22 crucial test results.