Learn More
Waters Corp Waters Symmetry C18 Column, 300Å, 5 μm, 4.6 mm X 150 mm
Catalog No. 50786032 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Symmetry C18 Column is a 300Å, 5 μm column (4.6 mm x 150 mm) for HPLC drug assays. It ensures long-term method compliance with consistent peak shape, selectivity, and reproducibility. Manufactured in cGMP, ISO9001, and ISO13485 certified facilities, it uses high purity raw materials, a monofunctionally bonded C18 ligand, and base-deactivated silica.
  • Sets the standard for HPLC drug assays.
  • Ensures long-term compliance of HPLC methods.
  • Offers outstanding reproducibility year on year, eliminating continuous method revalidation.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities.
  • Features a monofunctionally bonded C18 ligand and high purity base-deactivated silica.
  • Supplied with a three-page certificate of analysis.
Catalog No. 50-786-032 Supplier Waters Corp Supplier No. WAT106157
The Symmetry C18 Column is a 300Å, 5 μm column (4.6 mm x 150 mm) for HPLC drug assays. It ensures long-term method compliance with consistent peak shape, selectivity, and reproducibility. Manufactured in cGMP, ISO9001, and ISO13485 certified facilities, it uses high purity raw materials, a monofunctionally bonded C18 ligand, and base-deactivated silica.
  • Sets the standard for HPLC drug assays.
  • Ensures long-term compliance of HPLC methods.
  • Offers outstanding reproducibility year on year, eliminating continuous method revalidation.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities.
  • Features a monofunctionally bonded C18 ligand and high purity base-deactivated silica.
  • Supplied with a three-page certificate of analysis.