Learn More
Waters Corp Symmetry300 C18 Guard Column, 3.9 x 20 mm, 5 μm, 2-pack
Catalog No. 50780121 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry300 C18 Guard Columns are designed to protect analytical columns from chemical and particulate contamination in samples and mobile phases. This extends the lifetime of the analytical column and maintains its performance.
  • Protects analytical columns from chemical contamination.
  • Protects analytical columns from particulate contamination.
  • Extends the lifetime of the analytical column.
  • Maintains analytical column performance.
  • C18 material.
  • 3.9 x 20 mm dimensions.
  • 5 μm particle size.
  • Supplied as a 2-pack.
Catalog No. 50-780-121 Supplier Waters Corp Supplier No. 186000198
Symmetry300 C18 Guard Columns are designed to protect analytical columns from chemical and particulate contamination in samples and mobile phases. This extends the lifetime of the analytical column and maintains its performance.
  • Protects analytical columns from chemical contamination.
  • Protects analytical columns from particulate contamination.
  • Extends the lifetime of the analytical column.
  • Maintains analytical column performance.
  • C18 material.
  • 3.9 x 20 mm dimensions.
  • 5 μm particle size.
  • Supplied as a 2-pack.