Learn More
Waters Corp Waters Symmetry C4 Column, 300 Å, 3.5 μm, 4.6 mm, 150 mm
Supplier: Waters Corp 186000283
This Symmetry C4 Column supports legacy methods, ensuring long-term HPLC compliance. It offers consistent peak shape, selectivity, and reproducibility. Excelling in separating diverse peptides, its wide-pored, reversed-phase C4 chemistry is popular for large peptides and proteins.
- Ensures long-term HPLC method compliance
- Consistent peak shape, selectivity, and reproducibility
- Improved peak shape and capacity for peptide separation
- Wide-pored, reversed-phase C4 chemistry
- Ultrapure silica manufacturing, low silanol activity
- Chemistry: C4
- Separation mode: Reversed Phase
- Particle size: 3.5 μm
- Pore size: 300 Å
- Inner diameter: 4.6 mm
- Length: 150 mm
- pH range: 2 to 8 pH
- Maximum pressure: 6000 psi (415 Bar)
- Molecular weight range: 1000 to 30000
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.