Learn More
Waters Corp Waters Column, C4, 300 Å, 3.5 μm, 4.6 mm x 75 mm
Catalog No. 50780168 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column provides consistent peak shape, selectivity, and reproducibility for HPLC methods. It separates physiologically active peptides and is suitable for large peptides and proteins due to its C4 chemistry. It supports validation compliance for impurity and stability assays.
  • Supports legacy HPLC methods and long-term compliance.
  • Offers enhanced peak shape, selectivity, and reproducibility.
  • Separates various physiologically active peptides.
  • Ultrapure silica construction for low silanol activity.
  • Available in diverse particle sizes, chemistries, and dimensions.
  • Provides sharp peaks for biomolecules and increased peak capacity.
  • Mass spectrometry compatible (high- and low-mass situations).
  • Ensures validation compliance and accurate quantitation.
Catalog No. 50-780-168 Supplier Waters Corp Supplier No. 186000281
Add to Cart
Add to Cart
This column provides consistent peak shape, selectivity, and reproducibility for HPLC methods. It separates physiologically active peptides and is suitable for large peptides and proteins due to its C4 chemistry. It supports validation compliance for impurity and stability assays.
  • Supports legacy HPLC methods and long-term compliance.
  • Offers enhanced peak shape, selectivity, and reproducibility.
  • Separates various physiologically active peptides.
  • Ultrapure silica construction for low silanol activity.
  • Available in diverse particle sizes, chemistries, and dimensions.
  • Provides sharp peaks for biomolecules and increased peak capacity.
  • Mass spectrometry compatible (high- and low-mass situations).
  • Ensures validation compliance and accurate quantitation.