Learn More
Waters Corp Waters Column, C4, 3.5 μm, 300 Å, 2.1 x 50 mm
Supplier:  Waters Corp 186000277
Encompass_Preferred
Symmetry C4 Columns provide industry-standard performance for HPLC drug assays, ensuring compliance with excellent peak shape and selectivity. They effectively separate various physiologically active peptides, performing well in high and low mass applications, including mass spectrometry with low TFA or 0.1% formic acid mobile phases.
  • Ensures long-term compliance for HPLC drug assays.
  • Improves peak shape and capacity for peptide separation.
  • Separates hydrophilic, hydrophobic, neutral, and basic peptides.
  • Ideal for high/low mass applications, including mass spectrometry.
  • Delivers sharp biomolecule peaks; increases capacity for tryptic maps.
  • Features ultrapure silica with very low silanol activity.
  • High efficiency, reproducibility enhance precision and quantitation.
  • Wide-pored, reversed-phase C4 chemistry suits large peptides/proteins.
Catalog No. 50-780-164