Learn More
Waters Corp Waters Column, C4, 3.5 μm, 300 Å, 4.6 mm x 50 mm
Supplier: Waters Corp 186000280
Symmetry columns are designed to provide confidence in the long-term compliance of HPLC methods. They ensure no compromise on peak shape, selectivity, or reproducibility. These columns are suitable for a variety of particle sizes, chemistries, and dimensions, primarily supporting legacy methods.
- C4 chemistry.
- Reversed Phase separation mode.
- Silica particle substrate.
- pH range of 2 to 8 pH.
- Maximum pressure of 6000 psi (415 Bar).
- Endcapped.
- Low silanol activity.
- Molecular weight range of 1000 to 30000.
- Spherical particle shape.
- Particle size of 3.5 μm.
- Pore size of 300 Å.
- HPLC system.
- USP classification L26.
- Inner diameter: 4.6 mm.
- Length: 50 mm.
- Carbon load: 2.8 %.
- Application: Protein.
- Units per package: 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.