Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation C4 Column, 300 Å, 5 μm

Catalog No. 50780172 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Symmetry columns are designed to provide confidence in the long-term compliance of HPLC methods. They ensure consistent peak shape, selectivity, column-to-column and batch-to-batch reproducibility, and other critical performance characteristics. This column primarily supports legacy methods.

  • Features C4 chemistry and reversed phase separation mode
  • Silica particle substrate with a spherical shape
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped with low silanol activity
  • Molecular weight range of 1000 to 30000
  • Pore size of 300 Å and particle size of 5 μm
  • Dimensions: 2.1 mm inner diameter and 150 mm length
  • 2.8 % carbon load
  • Suitable for protein applications
  • One unit per package

Catalog No. 50-780-172 Supplier Waters Corp Supplier No. 186000285
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Symmetry columns are designed to provide confidence in the long-term compliance of HPLC methods. They ensure consistent peak shape, selectivity, column-to-column and batch-to-batch reproducibility, and other critical performance characteristics. This column primarily supports legacy methods.

  • Features C4 chemistry and reversed phase separation mode
  • Silica particle substrate with a spherical shape
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped with low silanol activity
  • Molecular weight range of 1000 to 30000
  • Pore size of 300 Å and particle size of 5 μm
  • Dimensions: 2.1 mm inner diameter and 150 mm length
  • 2.8 % carbon load
  • Suitable for protein applications
  • One unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.