Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Sentry Guard Cartridge, C4, 300 Å, 3.5 μm, 2.1 mm, 10 mm

Catalog No. 50780162 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Symmetry columns primarily support legacy methods, offering long-term compliance for HPLC methods. They provide consistent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility. These cartridges are available in various particle sizes, chemistries, and dimensions.

  • C4 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Molecular weight range of 1000 to 30000
  • Spherical particle shape
  • Particle size of 3.5 μm
  • Pore size of 300 Å
  • Sentry guard cartridge format
  • Surface area of 110
  • HPLC system compatibility
  • USP classification L26
  • Inner diameter of 2.1 mm
  • Length of 10 mm
  • Carbon load of 2.8%
  • Application for protein
  • Two units per package

Catalog No. 50-780-162 Supplier Waters Corp Supplier No. 186000275
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Symmetry columns primarily support legacy methods, offering long-term compliance for HPLC methods. They provide consistent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility. These cartridges are available in various particle sizes, chemistries, and dimensions.

  • C4 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Molecular weight range of 1000 to 30000
  • Spherical particle shape
  • Particle size of 3.5 μm
  • Pore size of 300 Å
  • Sentry guard cartridge format
  • Surface area of 110
  • HPLC system compatibility
  • USP classification L26
  • Inner diameter of 2.1 mm
  • Length of 10 mm
  • Carbon load of 2.8%
  • Application for protein
  • Two units per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.