Learn More
Waters Corp Waters Sentry Guard Cartridge, C4, 5 μm, 300 Å
Catalog No. 50780171 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sentry Guard Cartridge supports legacy HPLC methods, ensuring long-term compliance and consistent performance. It protects columns, minimizing particulate matter and chemical pollutants, and features tool-free replacement.
  • Ensures confidence in the long-term compliance and consistent performance of HPLC methods.
  • Maintains excellent peak shape, selectivity, and column-to-column/batch-to-batch reproducibility.
  • Protects columns, extending lifespan by minimizing particulate matter and chemical pollutants.
  • Offers tool-free replacement.
  • Produces sharp peaks for biomolecules and provides additional peak capacity for complex tryptic maps.
  • Features ultrapure silica for very low silanol activity, high efficiency, and reproducibility.
  • Enables better precision for impurity and stability tests, accurate quantitation, and adaptability to challenging procedures.
Catalog No. 50-780-171 Supplier Waters Corp Supplier No. 186000284
The Sentry Guard Cartridge supports legacy HPLC methods, ensuring long-term compliance and consistent performance. It protects columns, minimizing particulate matter and chemical pollutants, and features tool-free replacement.
  • Ensures confidence in the long-term compliance and consistent performance of HPLC methods.
  • Maintains excellent peak shape, selectivity, and column-to-column/batch-to-batch reproducibility.
  • Protects columns, extending lifespan by minimizing particulate matter and chemical pollutants.
  • Offers tool-free replacement.
  • Produces sharp peaks for biomolecules and provides additional peak capacity for complex tryptic maps.
  • Features ultrapure silica for very low silanol activity, high efficiency, and reproducibility.
  • Enables better precision for impurity and stability tests, accurate quantitation, and adaptability to challenging procedures.