Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Silica C18 Prep Guard Cartridge, 100Å, 7 μm, 19 x 10 mm

Catalog No. 50819053 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Symmetry columns are designed to maintain high standards for HPLC drug assays, ensuring long-term compliance for HPLC methods. They aim to deliver consistent peak shape, selectivity, and reproducibility from column-to-column and batch-to-batch. These columns are available in various particle sizes, chemistries, and dimensions.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Max pressure: 6000 psi (415 Bar)
  • Endcapped
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 7 μm
  • Pore size: 100 Å
  • Format: Prep guard cartridge
  • Surface area: 335
  • System: HPLC preparative
  • Technique: LC/MS, LC
  • USP classification: L1
  • Inner diameter: 19 mm
  • Length: 10 mm
  • Carbon load: 19%
  • Units per package: 1

Catalog No. 50-819-053 Supplier Waters Corp Supplier No. 186000717
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Symmetry columns are designed to maintain high standards for HPLC drug assays, ensuring long-term compliance for HPLC methods. They aim to deliver consistent peak shape, selectivity, and reproducibility from column-to-column and batch-to-batch. These columns are available in various particle sizes, chemistries, and dimensions.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Max pressure: 6000 psi (415 Bar)
  • Endcapped
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 7 μm
  • Pore size: 100 Å
  • Format: Prep guard cartridge
  • Surface area: 335
  • System: HPLC preparative
  • Technique: LC/MS, LC
  • USP classification: L1
  • Inner diameter: 19 mm
  • Length: 10 mm
  • Carbon load: 19%
  • Units per package: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.