Learn More
Waters Corp Waters Prep Column, C18, 7 μm, 19 mm × 300 mm
Supplier: Waters Corp WAT066245
The Symmetry C18 Prep Column ensures consistent, high-quality performance for HPLC drug assays by delivering excellent peak shape, selectivity, and reproducibility. Featuring a monofunctionally bound C18 ligand and high-purity, base-deactivated silica, it maximizes analytical efficiency and precision for long-term compliance.
- C18 chemistry
- Reversed phase separation mode
- Silica particle substrate
- pH range of 2 to 8
- Temperature limits up to 45°C
- Maximum pressure of 6000 psi (415 Bar)
- Endcapped
- Low silanol activity
- Spherical particle shape
- 7 μm particle size
- 100 Å pore size
- 19 mm inner diameter
- 300 mm length
- 19% carbon load
- Manufactured in cGMP, ISO9001, and ISO13485 certified facilities
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.