Learn More
Waters Corp Waters Guard Cartridge, C18, 7 μm, 100 Å, 7.8 × 10 mm
Supplier: Waters Corp 186000713
Symmetry columns set the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods. They offer excellent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility.
- Ensures long-term HPLC method compliance with excellent peak shape, selectivity, and reproducibility.
- Chemistry: C18; Reversed phase separation; Silica particle substrate.
- pH range: 2 to 8; Temperature limits: 45 °C; Max pressure: 6000 psi (415 Bar).
- Endcapped, with low silanol activity and spherical particle shape.
- Particle size: 7 μm; Pore size: 100 Å; Surface area: 335.
- Prep guard cartridge for HPLC preparative, LC/MS, and LC techniques.
- USP classification: L1; Inner diameter: 7.8 mm; Length: 10 mm.
- Carbon load: 19 %; 1 unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.