Learn More
Waters Corp Waters Symmetry C18 Prep Column, 5 μm, 7.8 mm × 50 mm
Catalog No. 50780127 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry columns are designed to set the standard for HPLC drug assays, ensuring confidence in the long-term compliance of HPLC methods. They offer excellent peak shape, selectivity, column-to-column and batch-to-batch reproducibility, and other critical performance characteristics. These columns are available in various particle sizes, chemistries, and dimensions.
  • Sets the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods.
  • Provides excellent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility.
  • Available in a variety of particle sizes, chemistries, and dimensions.
Catalog No. 50-780-127 Supplier Waters Corp Supplier No. 186000208
Symmetry columns are designed to set the standard for HPLC drug assays, ensuring confidence in the long-term compliance of HPLC methods. They offer excellent peak shape, selectivity, column-to-column and batch-to-batch reproducibility, and other critical performance characteristics. These columns are available in various particle sizes, chemistries, and dimensions.
  • Sets the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods.
  • Provides excellent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility.
  • Available in a variety of particle sizes, chemistries, and dimensions.