Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Prep Column, C18, 7 μm, 19 mm x 150 mm

Catalog No. 50785875 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Symmetry columns are engineered for HPLC drug assays, ensuring long-term method compliance. They offer consistent peak shape, selectivity, and reproducibility (column-to-column and batch-to-batch), without compromising critical performance.

  • C18 chemistry for reliable separations.
  • Reversed phase separation mode with silica substrate.
  • pH range: 2 to 8.
  • Temperature limit: 45°C.
  • Maximum pressure: 6000 psi (415 Bar).
  • Endcapped with low silanol activity.
  • Spherical particle shape, 7 μm particle size.
  • Pore size: 100 Å.
  • Prep column format, 19 mm inner diameter, 150 mm length.
  • Carbon load: 19 %.
  • Designed for HPLC, LC, and LC/MS techniques.

Catalog No. 50-785-875 Supplier Waters Corp Supplier No. WAT066240
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Symmetry columns are engineered for HPLC drug assays, ensuring long-term method compliance. They offer consistent peak shape, selectivity, and reproducibility (column-to-column and batch-to-batch), without compromising critical performance.

  • C18 chemistry for reliable separations.
  • Reversed phase separation mode with silica substrate.
  • pH range: 2 to 8.
  • Temperature limit: 45°C.
  • Maximum pressure: 6000 psi (415 Bar).
  • Endcapped with low silanol activity.
  • Spherical particle shape, 7 μm particle size.
  • Pore size: 100 Å.
  • Prep column format, 19 mm inner diameter, 150 mm length.
  • Carbon load: 19 %.
  • Designed for HPLC, LC, and LC/MS techniques.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.