Learn More
Waters Corp Waters Symmetry Prep Column, C18, 7 μm, 7.8 mm x 150 mm
Supplier: Waters Corp WAT066288
Symmetry columns continue to set the standard for HPLC drug assays, providing confidence in the long-term compliance of HPLC methods. They eliminate the need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics.
- Chemistry: C18
- Separation mode: Reversed Phase
- Particle substrate: Silica
- pH range: 2 to 8 pH
- Temperature limits: 45°C
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped: Yes
- Silanol activity: Low
- Particle shape: Spherical
- Particle size: 7 μm
- Endfitting type: Waters
- Pore size: 100 Å
- Format: Prep Column
- Surface area: 335
- System: HPLC Preparative
- Technique: LC/MS, LC
- USP classification: L1
- Inner diameter: 7.8 mm
- Length: 150 mm
- Carbon load: 19%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.