Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Prep Guard Cartridge, C8, 100Å, 5 μm, 7.8 mm x 10 mm

Catalog No. 50780475 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Symmetry columns are designed to set the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods. They maintain excellent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility. These columns are available in various particle sizes, chemistries, and dimensions.

  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Pore size: 100 Å
  • Particle size: 5 μm
  • Inner diameter: 7.8 mm
  • Length: 10 mm
  • Carbon load: 12 %

Catalog No. 50-780-475 Supplier Waters Corp Supplier No. 186000712
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Symmetry columns are designed to set the standard for HPLC drug assays, ensuring long-term compliance of HPLC methods. They maintain excellent peak shape, selectivity, and column-to-column and batch-to-batch reproducibility. These columns are available in various particle sizes, chemistries, and dimensions.

  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Pore size: 100 Å
  • Particle size: 5 μm
  • Inner diameter: 7.8 mm
  • Length: 10 mm
  • Carbon load: 12 %

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.